Assay Details
| Gene Symbol: apx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112030.1 | NP_001105500.1 | 483 | 66 | ||
| GenBank mRNA | AY104492.1 | - | 497 | 66 | ||
| Z34934.1 | - | 483 | 66 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

apx2| Gene Symbol: apx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112030.1 | NP_001105500.1 | 483 | 66 | ||
| GenBank mRNA | AY104492.1 | - | 497 | 66 | ||
| Z34934.1 | - | 483 | 66 | |||