Assay Details
| Gene Symbol: LOC100281203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154122.1 | NP_001147594.1 | 1532 | 85 | ||
| GenBank mRNA | BT041844.1 | - | 1619 | 85 | ||
| EU955911.1 | - | 1532 | 85 | |||
More Information
Set Membership: |
|

LOC100281203
Genomic Map