Assay Details
| Gene Symbol: lkrsdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111403.1 | NP_001104873.1 | 288 | 97 | ||
| GenBank mRNA | AF003551.2 | - | 465 | 97 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lkrsdh1| Gene Symbol: lkrsdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111403.1 | NP_001104873.1 | 288 | 97 | ||
| GenBank mRNA | AF003551.2 | - | 465 | 97 | ||
Set Membership: |
|