Assay Details
| Gene Symbol: LOC542103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111736.1 | NP_001105206.1 | 2958 | 79 | ||
| GenBank mRNA | AY187719.1 | - | 2958 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC542103| Gene Symbol: LOC542103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111736.1 | NP_001105206.1 | 2958 | 79 | ||
| GenBank mRNA | AY187719.1 | - | 2958 | 79 | ||
Set Membership: |
|