Assay Details
| Gene Symbol: drh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111771.1 | NP_001105241.1 | 565 | 97 | ||
| GenBank mRNA | AY466159.1 | - | 565 | 97 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

drh1| Gene Symbol: drh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111771.1 | NP_001105241.1 | 565 | 97 | ||
| GenBank mRNA | AY466159.1 | - | 565 | 97 | ||