Assay Details
| Gene Symbol: drh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111771.1 | NP_001105241.1 | 1695 | 65 | ||
| GenBank mRNA | AY107918.1 | - | 134 | 65 | ||
| AY466159.1 | - | 1695 | 65 | |||
| Gene Symbol: LOC100383848 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001176478.1 | NP_001169949.1 | 65 | |||
| GenBank mRNA | BT068436.2 | - | 65 | |||
More Information
Set Membership: |
|

drh1
Genomic Map