Assay Details
| Gene Symbol: hdt103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111932.1 | NP_001105402.1 | 921 | 87 | ||
| GenBank mRNA | U82815.1 | - | 921 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

hdt103| Gene Symbol: hdt103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111932.1 | NP_001105402.1 | 921 | 87 | ||
| GenBank mRNA | U82815.1 | - | 921 | 87 | ||
Set Membership: |
|