Assay Details
| Gene Symbol: pco072546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112114.1 | NP_001105584.1 | 1350 | 101 | ||
| GenBank mRNA | M80912.1 | - | 1350 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pco072546| Gene Symbol: pco072546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112114.1 | NP_001105584.1 | 1350 | 101 | ||
| GenBank mRNA | M80912.1 | - | 1350 | 101 | ||
Set Membership: |
|