Assay Details
| Gene Symbol: pdi12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112292.1 | NP_001105762.1 | 352 | 94 | ||
| GenBank mRNA | AY739294.1 | - | 352 | 94 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

pdi12| Gene Symbol: pdi12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112292.1 | NP_001105762.1 | 352 | 94 | ||
| GenBank mRNA | AY739294.1 | - | 352 | 94 | ||