Assay Details
| Gene Symbol: lpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112320.1 | NP_001105790.1 | 1013 | 82 | ||
| GenBank mRNA | AY772410.1 | - | 1013 | 82 | ||
| BT033394.1 | - | 1223 | 82 | |||
| BT041512.1 | - | 1078 | 82 | |||
More Information
Set Membership: |
|

lpa3
Genomic Map