Assay Details
| Gene Symbol: afd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112359.1 | NP_001105829.1 | 1456 | 85 | ||
| GenBank mRNA | AY788900.1 | - | 1456 | 85 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

afd1| Gene Symbol: afd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112359.1 | NP_001105829.1 | 1456 | 85 | ||
| GenBank mRNA | AY788900.1 | - | 1456 | 85 | ||