Assay Details
| Gene Symbol: GRF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112555.1 | NP_001106025.1 | 1145 | 58 | ||
| GenBank mRNA | EF515845.1 | - | 1145 | 58 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRF6| Gene Symbol: GRF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112555.1 | NP_001106025.1 | 1145 | 58 | ||
| GenBank mRNA | EF515845.1 | - | 1145 | 58 | ||
Set Membership: |
|