Assay Details
Gene Symbol: umc1324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136786.1 | NP_001130258.1 | 458 | 74 | ||
GenBank mRNA | BT033421.1 | - | 458 | 74 | ||
BT068070.1 | - | 293 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
