Assay Details
| Gene Symbol: LOC100191936 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137360.1 | NP_001130832.1 | 778 | 62 | ||
| GenBank mRNA | BT034194.1 | - | 778 | 62 | ||
| BT067210.1 | - | 810 | 62 | |||
| EU959699.1 | - | 813 | 62 | |||
More Information
Set Membership: |
|

LOC100191936
Genomic Map