Assay Details
| Gene Symbol: Rbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001138111.1 | NP_001131583.1 | 407 | 86 | ||
| GenBank mRNA | BT035044.1 | - | 397 | 86 | ||
| EU955105.1 | - | 407 | 86 | |||
| EU961074.1 | - | 323 | 86 | |||
| GU984781.1 | - | 323 | 86 | |||
More Information
Set Membership: |
|

Rbx1
Genomic Map