Assay Details
| Gene Symbol: magi50422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112515.1 | NP_001105985.1 | 955 | 101 | ||
| GenBank mRNA | BT037934.1 | - | 1006 | 101 | ||
| EF406364.1 | - | 955 | 101 | |||
More Information
Set Membership: |
|

magi50422
Genomic Map