Assay Details
| Gene Symbol: por1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111983.1 | NP_001105453.1 | 1646 | 107 | ||
| GenBank mRNA | X73429.1 | - | 1646 | 107 | ||
| Gene Symbol: LOC100382343 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001175090.1 | NP_001168561.1 | 2403 | 108 | ||
| GenBank mRNA | BT063975.1 | - | 2403 | 108 | ||
More Information
Set Membership: |
|

por1
Genomic Map