Assay Details
| Gene Symbol: rpl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139101.1 | NP_001132573.1 | 747 | 93 | ||
| GenBank mRNA | BT036465.1 | - | 747 | 93 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rpl10| Gene Symbol: rpl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139101.1 | NP_001132573.1 | 747 | 93 | ||
| GenBank mRNA | BT036465.1 | - | 747 | 93 | ||
Set Membership: |
|