Assay Details
| Gene Symbol: LOC100285201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158095.1 | NP_001151567.1 | 550 | 99 | ||
| GenBank mRNA | EU966332.1 | - | 364 | 99 | ||
| EU971250.1 | - | 550 | 99 | |||
More Information
Set Membership: |
|

LOC100285201
Genomic Map