Assay Details
| Gene Symbol: LOC100191250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136684.1 | NP_001130156.1 | 1246 | 86 | ||
| GenBank mRNA | BT033288.1 | - | 1246 | 86 | ||
| BT084537.1 | - | 1372 | 86 | |||
More Information
Set Membership: |
|

LOC100191250
Genomic Map