accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3O

CAS RN 100-06-1

IUPAC Name: 1-(4-methoxyphenyl)ethan-1-one
Synonyms: P-methoxyacetophenone acetanisole
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: NTPLXRHDUXRPNE-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(C)=O
Boiling Point 
  • (3)
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (7)
Percent Purity 
  • (2)
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (7)
  • (2)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)