accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100-12-9

IUPAC Name: 1-ethyl-4-nitrobenzene
Synonyms: 4-ethylnitrobenzene
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: RESTWAHJFMZUIZ-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)