accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100-15-2

IUPAC Name: N-methyl-4-nitroaniline
Synonyms: P-nitro-N-methylaniline
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: XIFJZJPMHNUGRA-UHFFFAOYSA-N
SMILES: CNC1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)