accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100-20-9

IUPAC Name: benzene-1,4-dicarbonyl dichloride
Synonyms: terephthaloyl chloride
Molecular Weight (g/mol): 203.02
Molecular Formula: C8H4Cl2O2
InChi Key: LXEJRKJRKIFVNY-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(C=C1)C(Cl)=O
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)