accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100-23-2

IUPAC Name: N,N-dimethyl-4-nitroaniline
Synonyms: P-nitro-N,N-dimethylaniline
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: QJAIOCKFIORVFU-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)