accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNOO

CAS RN 100-27-6

IUPAC Name: 2-(4-nitrophenyl)ethan-1-ol
Synonyms: 2-(P-nitrophenyl)ethanol
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: IKMXRUOZUUKSON-UHFFFAOYSA-N
SMILES: OCCC1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (5)
  • (3)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)