accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100-29-8

IUPAC Name: 1-ethoxy-4-nitrobenzene
Synonyms: P-nitrophenetole
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: NWPKEYHUZKMWKJ-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results