accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2C

CAS RN 100-40-3

IUPAC Name: 4-ethenylcyclohex-1-ene
Synonyms: 4-vinylcyclohexene
Molecular Weight (g/mol): 108.18
Molecular Formula: C8H12
InChi Key: BBDKZWKEPDTENS-UHFFFAOYNA-N
SMILES: C=CC1CCC=CC1
Percent Purity 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)