accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2C

CAS RN 100-42-5

IUPAC Name: ethenylbenzene
Synonyms: styrene fema number 3234 bulstren K-525-19 styron styrol styreen styrole ccris 564 styropol SO styrolene
Molecular Weight (g/mol): 104.15
Molecular Formula: C8H8
InChi Key: PPBRXRYQALVLMV-UHFFFAOYSA-N
SMILES: C=CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (15)
  • (6)
Quantity 
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (12)
  • (5)
Percent Purity 
  • (1)
  • (2)
  • (6)
  • (5)
Physical Form 
  • (11)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (1)
  • (2)
  • (12)
  • (15)
  • (6)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (12)
  • (5)
  • (1)
  • (2)
  • (6)
  • (5)
  • (11)
  • (2)
  • (4)
  • (3)
  • (1)