accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100-45-8

N

CAS RN 100-45-8

IUPAC Name: cyclohex-3-ene-1-carbonitrile
Synonyms: 3-cyclohexene-1-carbonitrile
Molecular Weight (g/mol): 107.16
Molecular Formula: C7H9N
InChi Key: GYBNBQFUPDFFQX-UHFFFAOYNA-N
SMILES: N#CC1CCC=CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)