accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClHg

CAS RN 100-56-1

IUPAC Name: chloro(phenyl)mercury
Synonyms: phenyl mercuric chloride
Molecular Weight (g/mol): 313.15
Molecular Formula: C6H5ClHg
InChi Key: AWGTVRDHKJQFAX-UHFFFAOYSA-M
SMILES: Cl[Hg]C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results