accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100-68-5

IUPAC Name: (methylsulfanyl)benzene
Synonyms: thioanisole
Molecular Weight (g/mol): 124.2
Molecular Formula: C7H8S
InChi Key: HNKJADCVZUBCPG-UHFFFAOYSA-N
SMILES: CSC1=CC=CC=C1
Boiling Point 
  • (6)
  • (4)
  • (4)
Molecular Weight (g/mol) 
  • (3)
  • (7)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (3)
  • (8)
  • (6)
  • (4)
  • (4)
  • (3)
  • (7)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (8)
  • (7)