accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 100-76-5

IUPAC Name: 1-azabicyclo[2.2.2]octane
Synonyms: quinuclidine 1,4-ethylenepiperidine chinuclidin
Molecular Weight (g/mol): 111.19
Molecular Formula: C7H13N
InChi Key: SBYHFKPVCBCYGV-UHFFFAOYSA-N
SMILES: C1CN2CCC1CC2
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)