accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3AlH3CCH3H3CCH3

CAS RN 100-99-2

IUPAC Name: tris(2-methylpropyl)alumane
Synonyms: triisobutylaluminium
Molecular Weight (g/mol): 198.33
Molecular Formula: C12H27Al
InChi Key: MCULRUJILOGHCJ-UHFFFAOYSA-N
SMILES: CC(C)C[Al](CC(C)C)CC(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)