accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiCH3CH3Cl

CAS RN 1000-50-6

IUPAC Name: butyl(chloro)dimethylsilane
Synonyms: silane, butylchlorodimethyl-
Molecular Weight (g/mol): 150.72
Molecular Formula: C6H15ClSi
InChi Key: MXOSTENCGSDMRE-UHFFFAOYSA-N
SMILES: CCCC[Si](C)(C)Cl
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)