accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3NC(?)NSiH3CH3CCH3

CAS RN 1000-70-0

IUPAC Name: 2,2,6,6-tetramethyl-3,5-diaza-2,6-disilahepta-3,4-diene
Synonyms: 2,2,6,6-tetramethyl-3,5-diaza-2,6-disilahepta-3,4-diene
Molecular Weight (g/mol): 186.41
Molecular Formula: C7H18N2Si2
InChi Key: KSVMTHKYDGMXFJ-UHFFFAOYSA-N
SMILES: C[Si](C)(C)N=C=N[Si](C)(C)C
Boiling Point 
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)