accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1000-86-8

IUPAC Name: 2,4-dimethylpenta-1,3-diene
Synonyms: 1,3-pentadiene, 2,4-dimethyl-
Molecular Weight (g/mol): 96.17
Molecular Formula: C7H12
InChi Key: CMSUNVGIWAFNBG-UHFFFAOYSA-N
SMILES: CC(C)=CC(C)=C
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)