accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrNOOHNNH2HCl

CAS RN 100032-79-9

IUPAC Name: (4-bromo-2-nitrophenyl)hydrazine hydrochloride
Synonyms: (4-bromo-2-nitrophenyl)hydrazine hydrochloride
Molecular Weight (g/mol): 268.5
Molecular Formula: C6H7BrClN3O2
InChi Key: SIBURLORIWZQNT-UHFFFAOYSA-N
SMILES: Cl.NNC1=C(C=C(Br)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results