accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1000342-11-9

IUPAC Name: methyl 3-amino-5-bromo-2-methylbenzoate
Synonyms: methyl 3-amino-5-bromo-2-methylbenzoate
Molecular Weight (g/mol): 244.09
Molecular Formula: C9H10BrNO2
InChi Key: NMLOSXSDLWFBKT-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=CC(N)=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results