accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHNN

CAS RN 1000576-51-1

IUPAC Name: methyl 3-cyano-1H-indole-6-carboxylate
Synonyms: methyl 3-cyano-1H-indole-6-carboxylate
Molecular Weight (g/mol): 200.2
Molecular Formula: C11H8N2O2
InChi Key: YBZVEPCMUWGFNW-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C2C(NC=C2C#N)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results