accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10008-73-8

IUPAC Name: 5-methylideneoxolan-2-one
Synonyms: α'-angelica lactone
Molecular Weight (g/mol): 98.1
Molecular Formula: C5H6O2
InChi Key: SIFBVNDLLGPEKT-UHFFFAOYSA-N
SMILES: C=C1CCC(=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results