accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrFF

CAS RN 1000994-95-5

IUPAC Name: 1-bromo-4-(1,1-difluoroethyl)benzene
Synonyms: 1-bromo-4-(1,1-difluoroethyl)benzene
Molecular Weight (g/mol): 221.05
Molecular Formula: C8H7BrF2
InChi Key: QXIBKCFAFRHORF-UHFFFAOYSA-N
SMILES: CC(F)(F)C1=CC=C(Br)C=C1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)