accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1001-26-9

IUPAC Name: ethyl (2E)-3-ethoxyprop-2-enoate
Synonyms: ethyl 3-ethoxy-2-propenoate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: ITQFPVUDTFABDH-AATRIKPKSA-N
SMILES: CCO\C=C\C(=O)OCC
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (1)