accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1001-53-2

IUPAC Name: N-(2-aminoethyl)acetamide
Synonyms: N-(2-aminoethyl)acetamide
Molecular Weight (g/mol): 102.14
Molecular Formula: C4H10N2O
InChi Key: DAKZISABEDGGSV-UHFFFAOYSA-N
SMILES: CC(=O)NCCN
Boiling Point 
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)