accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CCH3OONNNNOO

CAS RN 1001067-09-9

IUPAC Name: 2-(trimethylsilyl)ethyl 3-nitro-1H-1,2,4-triazole-1-carboxylate
Synonyms: 2-(trimethylsilyl)ethyl 3-nitro-1,2,4-triazole-1-carboxylate
Molecular Weight (g/mol): 258.31
Molecular Formula: C8H14N4O4Si
InChi Key: BHTJYIDOVYPNKR-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CCOC(=O)N1C=NC(=N1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results