accessibility menu, dialog, popup

UserName

Viewing profile as user:
HHBrBrH2NNH2NSNH2NH

CAS RN 100130-32-3

IUPAC Name: dihydrogen ({2-[(diaminomethylidene)amino]ethyl}sulfanyl)methanimidamide dibromide
Synonyms: dihydrogen {2-[(diaminomethylidene)amino]ethyl}sulfanylmethanimidamide dibromide
Molecular Weight (g/mol): 323.05
Molecular Formula: C4H13Br2N5S
InChi Key: GPWJSTKHQMIXCA-UHFFFAOYSA-N
SMILES: [H+].[H+].[Br-].[Br-].NC(N)=NCCSC(N)=N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results