accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPSOCH3ONNH3CCH3CH3

CAS RN 100155-47-3

IUPAC Name: O,O-diethyl O-6-methyl-2-(propan-2-yl)pyrimidin-4-yl phosphorothioate
Synonyms: diazinon caswell no. 342 nucidol dazzel kleen-dok basudin 10 G dimpylate drawizon exodin disonex
Molecular Weight (g/mol): 304.34
Molecular Formula: C12H21N2O3PS
InChi Key: FHIVAFMUCKRCQO-UHFFFAOYSA-N
SMILES: CCOP(=S)(OCC)OC1=NC(=NC(C)=C1)C(C)C
Grade 
  • (1)
Industry Type 
  • (1)
Percent Purity 
  • (1)
Technique 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)