accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10017-11-5

IUPAC Name: hydrogen prop-2-en-1-amine chloride
Synonyms: hydrogen allylamine chloride
Molecular Weight (g/mol): 93.55
Molecular Formula: C3H8ClN
InChi Key: MLGWTHRHHANFCC-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC=C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results