accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10017-53-5

IUPAC Name: 2-(3-methylphenoxy)acetamide
Synonyms: 2-(3-methylphenoxy)acetamide
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: GOVBTAZVUFGTFB-UHFFFAOYSA-N
SMILES: CC1=CC(OCC(N)=O)=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results