accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10017-56-8

IUPAC Name: 2-[bis(2-hydroxyethyl)amino]ethan-1-ol; phosphoric acid
Synonyms: phosphoric acid; triethanolamine
Molecular Weight (g/mol): 247.18
Molecular Formula: C6H18NO7P
InChi Key: NHFDKKSSQWCEES-UHFFFAOYSA-N
SMILES: OP(O)(O)=O.OCCN(CCO)CCO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results